Learn More
Fisherbrand™ Inkless Chart Recorders for Isotemp Ultra-Low Temperature Chest Freezers
Product Code. 17838020 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17838020 Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer Each
17808010 Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17838020
Record seven days of temperature performance at temperatures to -150°C using Fisherbrand™ Isotemp Chart Recorders for Ultra-Low Temperature Freezers.
  • Seven-day recorder
  • Functions to -150°C
  • Factory installed
TRUSTED_SUSTAINABILITY
Specifications
Description Factory Installed
Type Chart Recorder
Temperature Range -150°C
For Use With (Equipment) Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
Static Holding Time 7 Days