Learn More
Agilent BioTek Injector Accessory
GREENER_CHOICE
Product Code. 16562132 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
BT8040036
BT8040037
Unit Size:
Each
Product Code. 16562132
Brand: Agilent BioTek 1340518
Injector Accessory is suitable for a Dual Reagent Injector and features feedthrough assembly with a straight dispense tip.
  • Accessory for Dual Reagent Injector Module
  • Includes straight injector tips
  • Includes feedthrough assembly
TRUSTED_SUSTAINABILITY
Specifications
Product Type Injector Accessory
For Use With (Equipment) BT8040037