missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin beta B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17577-1-AP-20UL
This item is not returnable.
View return policy
Description
The inhibin beta B subunit joins the alpha subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumour-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. Because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. Furthermore, the beta B subunit forms a homodimer, activin B, and also joins with the beta A subunit to form a heterodimer, activin AB, both of which stimulate FSH secretion.Specifications
| Inhibin beta B | |
| Polyclonal | |
| Unconjugated | |
| INHBB | |
| Activin beta B chain, INHBB, Inhibin beta B chain, inhibin, beta B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16324, 25196, 3625 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09529, P17491, Q04999 | |
| INHBB | |
| Inhibin beta B Fusion Protein Ag10956 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction