missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INHAT-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£167.00 - £310.00
Specifications
| Antigen | INHAT-1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18039965
|
Novus Biologicals
NB100-56353SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18700684
|
Novus Biologicals
NB100-56353 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INHAT-1 Polyclonal specifically detects INHAT-1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| INHAT-1 | |
| Western Blot 1-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.1 mg/ml BSA with 0.02% Sodium Azide | |
| This antibody was raised against synthetic peptides of amino acids 66-81 and 135-151 of human INHAT-1. Sequence homology is 100% in mouse and rat. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| TAF-1a, TAF-1b | |
| IgG | |
| Protein G purified | |
| An approx. 37 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title