missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INHAT-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56353
This item is not returnable.
View return policy
Description
INHAT-1 Polyclonal specifically detects INHAT-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| INHAT-1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.1 mg/ml BSA with 0.02% Sodium Azide | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| An approx. 37 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-2 ug/ml | |
| TAF-1a, TAF-1b | |
| This antibody was raised against synthetic peptides of amino acids 66-81 and 135-151 of human INHAT-1. Sequence homology is 100% in mouse and rat. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction