missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INHA Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA550841

 View more versions of this product

Product Code. 17910913

  • £333.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INHA
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P05111, P17490
INHA
Recombinant Human INHa protein, Ala25-Cys363 (Accession #P05111).
200 μL
Primary
Human, Rat
Antibody
IgG2a κ
Western Blot
C1
Unconjugated
INHA
A inhibin subunit; A-inhibin subunit; AW555078; IHA; Inha; inhibin; inhibin alpha; inhibin alpha chain; inhibin alpha subunit
Mouse
Protein A/G
RUO
24504, 3623
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.