missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INHA Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £333.00
Specifications
| Antigen | INHA |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17988742
|
Invitrogen™
MA550842 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17910913
![]() |
Invitrogen™
MA550841 |
200 μL |
£333.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone.Specifications
| INHA | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| A inhibin subunit; A-inhibin subunit; AW555078; IHA; Inha; inhibin; inhibin alpha; inhibin alpha chain; inhibin alpha subunit | |
| INHA | |
| IgG2a κ | |
| Protein A/G | |
| INHA |
| C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P05111, P17490 | |
| 24504, 3623 | |
| Recombinant Human INHa protein, Ala25-Cys363 (Accession #P05111). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title