missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16187-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specifically against p47ING3.
The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This gene can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of this gene has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of this gene were detected in head and neck cancers. Two alternatively spliced transcript variants encoding different isoforms have been observed.Specifications
| ING3 | |
| Polyclonal | |
| Unconjugated | |
| ING3 | |
| Eaf4, ING2, ING3, Inhibitor of growth protein 3, p47ING3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 312154, 54556, 71777 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q498T3, Q8VEK6, Q9NXR8 | |
| Ing3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction