missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING2 Mouse anti-Human, Mouse, Rat, Clone: 2C3A7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ING2 |
|---|---|
| Clone | 2C3A7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868455
|
Proteintech
66512-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858455
|
Proteintech
66512-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.Specifications
| ING2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ING1L, ING1Lp, ING2, Inhibitor of growth protein 2, p32, p33ING2 | |
| Ing2 | |
| IgG1 | |
| Protein A | |
| Antibody |
| 2C3A7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Human | |
| Q9ESK4, Q9H160 | |
| 290744, 3622, 69260 | |
| ING2 Fusion Protein Ag19291 | |
| Primary | |
| -20°C | |
| ING2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title