missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING2 Mouse anti-Human, Mouse, Rat, Clone: 2C3A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66512-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.Specifications
| ING2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ESK4, Q9H160 | |
| Ing2 | |
| ING2 Fusion Protein Ag19291 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2C3A7 | |
| Unconjugated | |
| ING2 | |
| ING1L, ING1Lp, ING2, Inhibitor of growth protein 2, p32, p33ING2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 290744, 3622, 69260 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction