missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A Virus H1N1 Neuraminidase Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
£170.00 - £506.00
Specifications
| Antigen | Neuraminidase |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757337
![]() |
R&D Systems
AF4858-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18051862
|
R&D Systems
AF4858 |
100 μg |
£506.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuraminidase Polyclonal specifically detects Neuraminidase in Influenza A Virus H1N1 samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Neuraminidase | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| Influenza A virus H1N1 | |
| NANH, Sialidase | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| AAF77036 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant Influenza A Virus H1N1 Neuraminidase Ser37-Lys469 Accession # AAF77036 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects Influenza A Virus H1N1 Neuraminidase in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title