missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A Virus H1N1 Neuraminidase Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4858-SP
This item is not returnable.
View return policy
Description
Neuraminidase Polyclonal specifically detects Neuraminidase in Influenza A Virus H1N1 samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Neuraminidase | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NANH, Sialidase | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant Influenza A Virus H1N1 Neuraminidase Ser37-Lys469 Accession # AAF77036 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAF77036 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| Detects Influenza A Virus H1N1 Neuraminidase in direct ELISAs and Western blots. | |
| Influenza A virus H1N1 | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction