missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H7N7 Hemagglutinin Rabbit anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Antibody pair for Solid phase sandwich ELISA
£342.00 - £832.00
Specifications
| Antigen | Influenza A H7N7 Hemagglutinin |
|---|---|
| Applications | ELISA |
| Conjugate | HRP |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359253
|
Novus Biologicals
NBP3-06704-5PLATES |
5 Plates |
£342.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18372412
|
Bio-Techne
NBP3-06704-15PLATES |
15 Plates |
£832.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Solid phase sandwich ELISA for quantitative determination of Influenza A H7N7 HemagglutininSpezifikation
| Influenza A H7N7 Hemagglutinin | |
| HRP | |
| RUO | |
| Storage of components varies. See protocol for specific instructions. |
| ELISA | |
| Rabbit | |
| Virus | |
| Antibody Pairs |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts