missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H7N7 Hemagglutinin Rabbit anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Description
Solid phase sandwich ELISA for quantitative determination of Influenza A H7N7 Hemagglutinin
Specifications
Specifications
| Antigen | Influenza A H7N7 Hemagglutinin |
| Applications | ELISA |
| Conjugate | HRP |
| Host Species | Rabbit |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Target Species | Virus |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Product Type | Antibody Pairs |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?