missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin anti-Influenza virus, Antibody Pair, Novus Biologicals™
Antibody pair for Solid phase sandwich ELISA
£349.00 - £832.00
Specifications
| Antigen | Influenza A H5N1 Hemagglutinin |
|---|---|
| Applications | ELISA |
| Conjugate | Unconjugated |
| Regulatory Status | RUO |
| Research Discipline | Immunology, Virology Bacteria and Parasites |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18338105
|
Novus Biologicals
NBP3-12818-5PLATES |
5 Plates |
£349.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18370147
|
Novus Biologicals
NBP3-12818-15PLATES |
15 Plates |
£832.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Solid phase sandwich ELISA for quantitative determination of Influenza A H5N1 HemagglutininSpecifications
| Influenza A H5N1 Hemagglutinin | |
| Unconjugated | |
| Immunology, Virology Bacteria and Parasites | |
| Storage of components varies. See protocol for specific instructions. |
| ELISA | |
| RUO | |
| Influenza virus | |
| Antibody Pairs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title