missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin anti-Influenza virus, Antibody Pair, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Solid phase sandwich ELISA for quantitative determination of Influenza A H5N1 Hemagglutinin
Specifications
Specifications
| Antigen | Influenza A H5N1 Hemagglutinin |
| Applications | ELISA |
| Conjugate | Unconjugated |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Research Discipline | Immunology, Virology Bacteria and Parasites |
| Target Species | Influenza virus |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Product Type | Antibody Pairs |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?