missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Nucleoprotein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£181.00 - £417.00
Specifications
| Antigen | Influenza A H1N1 Nucleoprotein |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-3 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 24086762), Immunohistochemistry-Paraffin reported in scientific literature (PMID 16873262) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18049993
|
Novus Biologicals
NB100-56570SS |
0.025 mg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795013
|
Novus Biologicals
NB100-56570 |
0.1 mg |
£417.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza A H1N1 Nucleoprotein Polyclonal specifically detects Influenza A H1N1 Nucleoprotein in Human, Mouse, Influenza A Virus H1N1, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Influenza A H1N1 Nucleoprotein | |
| Western Blot 1-3 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 24086762), Immunohistochemistry-Paraffin reported in scientific literature (PMID 16873262) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Influenza virus, Virus | |
| NP_001081025.1 | |
| IgG | |
| Protein G purified | |
| Influenza A virus (A/Puerto Rico/8/34/Mount Sinai (H1N1) segment 5) nuclear protein. |
| 1.0 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS with 0.05% Sodium Azide | |
| Recombinant Influenza A H1N1 Nucleoprotein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title