missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Nucleoprotein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-56570SS
This item is not returnable.
View return policy
Description
Influenza A H1N1 Nucleoprotein Polyclonal specifically detects Influenza A H1N1 Nucleoprotein in Human, Mouse, Influenza A Virus H1N1, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Influenza A H1N1 Nucleoprotein | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Influenza virus, Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 1-3 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 24086762), Immunohistochemistry-Paraffin reported in scientific literature (PMID 16873262) | |
| NP_001081025.1 | |
| Recombinant Influenza A H1N1 Nucleoprotein. | |
| 0.025 mg | |
| Immunology | |
| Influenza A virus (A/Puerto Rico/8/34/Mount Sinai (H1N1) segment 5) nuclear protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction