missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M1 Antibody (02) - (A/Puerto Rico/8/34/Mount Sinai), Novus Biologicals™
Mouse Monoclonal Antibody
£190.00 - £348.00
Specifications
| Antigen | Influenza A H1N1 M1 |
|---|---|
| Clone | 02 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656016
|
Novus Biologicals
NBP3-06633-100ul |
100 μg |
£348.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654706
|
Novus Biologicals
NBP3-06633-50ul |
50 μg |
£190.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza A H1N1 M1 Monoclonal antibody specifically detects Influenza A H1N1 M1 in Influenza A virus H1N1 samples. It is validated for ELISASpecifications
| Influenza A H1N1 M1 | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG1 | |
| Protein A purified |
| 02 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Influenza A virus H1N1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H1N1 (Accession#: NP_040978.1; Ser 2-Lys 252). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title