missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M1 Antibody (02) - (A/Puerto Rico/8/34/Mount Sinai), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06633-50ul
This item is not returnable.
View return policy
Description
Influenza A H1N1 M1 Monoclonal antibody specifically detects Influenza A H1N1 M1 in Influenza A virus H1N1 samples. It is validated for ELISA
Specifications
| Influenza A H1N1 M1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H1N1 (Accession#: NP_040978.1; Ser 2-Lys 252). | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| 02 | |
| ELISA 1:1000-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza A virus H1N1 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction