missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INF2 Mouse anti-Human, CoraLite 594, Clone: 4A11A7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | INF2 |
|---|---|
| Clone | 4A11A7 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17761439
|
Proteintech
CL594-66910-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17669966
|
Proteintech
CL594-66910-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INF2 Monoclonal antibody apecifically detetects INF2 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| INF2 | |
| Immunofluorescence | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 64423 | |
| INF2 fusion protein Ag17043 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 4A11A7 | |
| Monoclonal | |
| Liquid | |
| Human | |
| BC008756 | |
| INF2 | |
| IgG2a | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title