missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INF2 Mouse anti-Human, CoraLite 594, Clone: 4A11A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66910-20UL
This item is not returnable.
View return policy
Description
INF2 Monoclonal antibody apecifically detetects INF2 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| INF2 | |
| Monoclonal | |
| 1000 ug/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| INF2 | |
| INF2 fusion protein Ag17043 | |
| 20 μL | |
| 64423 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG2a |
| Immunofluorescence | |
| 4A11A7 | |
| CoraLite 594 | |
| BC008756 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction