missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde-13C, TRC

Product Code. 30133314
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133314 50 mg 50mg
17962065 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133314 Supplier Toronto Research Chemicals Supplier No. TRCI61499250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093452-52-8
Molecular Formula C813CH7NO
Molecular Weight (g/mol) 146.15
Synonym 3-Formylindole-13C, 2H-Indole-3-carboxaldehyde-13C;
IUPAC Name 1H-indole-3-carbaldehyde-13C
SMILES [H][13C](C1=CNC2=CC=CC=C21)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H7NO/c11-6-7-5-10-9-4-2-1-3-8(7)9/h1-6,10H/i6+1
Formula Weight 146.06
Percent Purity >95%
Chemical Name or Material Indole-3-carboxaldehyde-13C
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.