missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Indo-1 AM Calcium Sensor Dye (UltraPure Grade)
Click to view available options
Quantity:
20 x 50 μg
Unit Size:
Pack of 20
Description
- Unbound Indo-1 has a peak emission at 475nm
- Upon binding calcium, peak emission shifts down to 410nm
- In flow cytometry this can be measured over time and can be represented as a ratio of the two emission wavelengths
- To optimize the ratio between the two emissions, unbound Indo-1 fluorescence should be collected using a filter above 475nm; bound Indo-1 fluorescence should be collected using a filter below 400nm
- Because the emission profile of Indo-1 is broad, multicolor flow analysis using eFluor ™ 450, eVolve™ 605 or eVolve™ 655 is not possible, however fluorochrome conjugated antibodies utilizing the 488nm or 633 nm are quite compatible with Indo-1
- Molecular weight: 1009.91g/mol
- Peak excitation: 346nm
- Peak emission: 475nm
- Reconstitute in high-quality, freshly opened DMSO
- Recommended concentration: 1 mM
- Once reconstituted, store at −20°C with dessicant and use within a short period of time. Avoid freeze-thawing
Specifications
Specifications
| Color | Blue |
| Detection Method | Fluorescence |
| For Use With (Application) | Immunohistochemistry (IHC), Flow Cytometry |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Dye |
| Emission | 346/475 |
| Product Line | eBioscience |
| Quantity | 20 x 50 μg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction