missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) hydroxide, 99.998% (metals basis)

Product Code. 11311928
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311928 10 g 10g
11301928 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311928 Supplier Thermo Scientific Alfa Aesar Supplier No. 011855.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20661-21-6

Indium(III) hydroxide, acts as a precursor to indium(III) oxide. It is used in batteries, glasses, ceramics, chemical reagents, semiconductor materials. It is also used used for manufacturing of panel displays and demist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20661-21-6
MDL Number MFCD00011056

Specifications

Melting Point 150°C (in -H2O)
Physical Form Powder
Quantity 10 g
Solubility Information It is insoluble in water.
Formula Weight 165.84
Percent Purity 99.998%
Assay (metals basis)
Chemical Name or Material Indium(III) hydroxide
compliance-icons
Product Identifier
  • Indium hydroxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Indium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.