accessibility menu, dialog, popup

UserName

OH2InOH2H2O

CAS RN 20661-21-6

IUPAC Name: indiganetriol
Synonyms: indium(III) hydroxide
Molecular Weight (g/mol): 168.86
Molecular Formula: H6InO3
InChi Key: ZMFWDTJZHRDHNW-UHFFFAOYSA-N
SMILES: O.O.O.[In]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results