missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Indigo
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Product Type | Dye Reagents |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : < 1 mg/mL (insoluble or slightly soluble) ∣ H2O : < 0.1 mg/mL (insoluble) |
| Target | NO Synthase ∣ MMP ∣ Endogenous Metabolite |
| Molecular Weight (g/mol) | 262.26 g/mol |
| CAS | 482-89-3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?