missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indian Hedgehog/Ihh Antibody (9L16) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£161.00 - £437.00
Specifications
| Antigen | Indian Hedgehog/Ihh |
|---|---|
| Clone | 9L16 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18157002
|
Novus Biologicals
NBP2-22020-0.025MG |
0.025 mg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094015
|
Novus Biologicals
NBP2-22020 |
0.1 mg |
£437.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.Specifications
| Indian Hedgehog/Ihh | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Stem Cell Signaling Pathway | |
| NP_002172 | |
| 3549 | |
| IgG2 | |
| Protein G purified |
| 9L16 | |
| Western Blot 1:500-1:1000, ELISA 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
| IHH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title