missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indian Hedgehog/Ihh Antibody (9L16) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22020-0.025MG
This item is not returnable.
View return policy
Description
Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Indian Hedgehog/Ihh | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, ELISA 1:2000 | |
| NP_002172 | |
| IHH | |
| This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Indian Hedgehog Nterminus. Rat anti human Indian Hedgehog N-Term NP_002172. | |
| 0.025 mg | |
| Stem Cell Signaling Pathway | |
| 3549 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 9L16 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction