Learn More
Indapamide, Thermo Scientific™
£30.60 - £83.30
Chemical Identifiers
| CAS | 26807-65-8 |
|---|---|
| Molecular Formula | C16H16ClN3O3S |
| Molecular Weight (g/mol) | 365.83 |
| MDL Number | MFCD00079375 |
| InChI Key | NDDAHWYSQHTHNT-UHFFFAOYNA-N |
| Synonym | indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol |
| PubChem CID | 3702 |
| ChEBI | CHEBI:5893 |
| IUPAC Name | 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide |
| SMILES | CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15436009
|
Thermo Scientific Alfa Aesar
J63846.03 |
1g |
£31.30 £30.60 / 1g Save £0.70 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15446009
|
Thermo Scientific Alfa Aesar
J63846.06 |
5g |
£85.10 £83.30 / 5g Save £1.80 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Used as an antihypertensive
- Diuretic
Chemical Identifiers
| 26807-65-8 | |
| 365.83 | |
| NDDAHWYSQHTHNT-UHFFFAOYNA-N | |
| 3702 | |
| 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide |
| C16H16ClN3O3S | |
| MFCD00079375 | |
| indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol | |
| CHEBI:5893 | |
| CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O |
Specifications
| 26807-65-8 | |
| MFCD00079375 | |
| indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol | |
| NDDAHWYSQHTHNT-UHFFFAOYNA-N | |
| 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide | |
| 3702 | |
| 365.83 |
| C16H16ClN3O3S | |
| 14,4935 | |
| Soluble in ethanol; Insoluble in water | |
| CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O | |
| 365.83 | |
| CHEBI:5893 | |
| Indapamide |
Safety and Handling
missing translation for 'einecsNumber' : 248-012-7
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.