missing translation for 'onlineSavingsMsg'
Learn More

Indapamide, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63846.03

Quantity 1g

 View more versions of this product

Product Code. 15436009

  • ONLINE EXCLUSIVE PRICE
    £31.30
    £30.60 / 1g
    Save £0.70
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Used as an antihypertensive
  • Diuretic
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

26807-65-8
365.83
NDDAHWYSQHTHNT-UHFFFAOYNA-N
3702
4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
C16H16ClN3O3S
MFCD00079375
indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol
CHEBI:5893
CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O

Specifications

26807-65-8
MFCD00079375
14,4935
Soluble in ethanol; Insoluble in water
CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O
365.83
CHEBI:5893
Indapamide
C16H16ClN3O3S
1g
indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol
NDDAHWYSQHTHNT-UHFFFAOYNA-N
4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
3702
365.83
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.