missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX InCELL Hunter™ eXpress PIM1 Kinase Binding Assay
Description
The InCELL Hunter™ eXpress PIM1 Kinase Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with kinases. This kit assesses compound-engagement-based stabilization of PIM1 via detection of PIM1 levels. This product uses EFC technology. The cell line included overexpresses PIM1 target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | A549 |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | PIM1 |
| Cell Line | A549 |
| Target | PIM1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?