missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX InCELL Hunter™ eXpress CSF1R Kinase Binding Assay

Product Code. 17304799
Encompass
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17304799

Brand: DiscoverX 960047E15CP0M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The InCELL Hunter™ eXpress CSF1R Kinase Binding Assay measures target engagement via compound-induced stabilization of CSF1R target protein. eXpress kits contain all assay materials: cells, reagents, and plates.

The InCELL Hunter™ eXpress CSF1R Kinase Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with kinases. This kit assesses compound-engagement-based stabilization of CSF1R via detection of CSF1R levels. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry A549
For Use With (Application) Assay-Ready eXpress Kits
Quantity 200 dp (2 x 96-well)
Target Entry CSF1R
Cell Line A549
Target CSF1R
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.