missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£170.00 - £345.00
Specifications
| Antigen | Importin-9 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18046455
|
Novus Biologicals
NB100-56499SS |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079773
|
Novus Biologicals
NB100-56499 |
0.1 mg |
£345.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Importin-9 Polyclonal specifically detects Importin-9 in Human, Mouse, Rat, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Knockout Validated.Specifications
| Importin-9 | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine, Primate, Rhesus Macaque | |
| Q96P70 | |
| 55705 | |
| A portion of amino acids 850-950 of human Importin-9 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9 | |
| IPO9 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c), mouse (isoforms 1 and 2), rat, dog and rhesus monkey, 88% homologous in chicken, 83% homologous in opossum, and 78% in bovine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title