missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56499
This item is not returnable.
View return policy
Description
Importin-9 Polyclonal specifically detects Importin-9 in Human, Mouse, Rat, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Knockout Validated.
Specifications
| Importin-9 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c), mouse (isoforms 1 and 2), rat, dog and rhesus monkey, 88% homologous in chicken, 83% homologous in opossum, and 78% in bovine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| Q96P70 | |
| IPO9 | |
| A portion of amino acids 850-950 of human Importin-9 was used as the immunogen. | |
| 0.1 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55705 | |
| Human, Mouse, Rat, Canine, Primate, Rhesus Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction