missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPDH2 Recombinant Rabbit Monoclonal Antibody (23GB6670), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | IMPDH2 |
|---|---|
| Clone | 23GB6670 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089350
![]() |
Invitrogen™
MA553090 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087728
|
Invitrogen™
MA553091 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IMPDH2 Recombinant Monoclonal antibody specifically detects IMPDH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| IMPDH2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| E9PU28, P12268, P24547 | |
| 23918, 301005, 3615 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6670 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| HAMAP-Rule:MF_03156}; IMP (inosine 5'-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP dehydrogenase 2 {ECO:0000255; IMP dehydrogenase type II; IMP oxireductase 2; IMPD; IMPD 2; IMPD 2 {ECO:0000255; IMPD2; IMPDH 2; IMPDH 2 {ECO:0000255; IMPDH II; Impdh2; IMPDH-II; inosine 5' phosphate dehydrogenase 2; inosine 5-monophosphate dehydrogenase 2; inosine 5'-phosphate dehydrogenase 2; inosine monophosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; inosine-5'-monophosphate dehydrogenase 2; inosine-5'-monophosphate dehydrogenase 2 {ECO:0000255 | |
| A synthesized peptide derived from human IMPDH2 (120-175AA). | |
| Primary | |
| -20°C | |
| IMPDH2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title