missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPDH2 Recombinant Rabbit Monoclonal Antibody (23GB6670), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553090
This item is not returnable.
View return policy
Description
This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5'-monophosphate into xanthine-5'-monophosphate, which is then converted into guanosine-5'-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in malignant transformation.
Specifications
| IMPDH2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IMPDH2 | |
| HAMAP-Rule:MF_03156}; IMP (inosine 5'-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP dehydrogenase 2 {ECO:0000255; IMP dehydrogenase type II; IMP oxireductase 2; IMPD; IMPD 2; IMPD 2 {ECO:0000255; IMPD2; IMPDH 2; IMPDH 2 {ECO:0000255; IMPDH II; Impdh2; IMPDH-II; inosine 5' phosphate dehydrogenase 2; inosine 5-monophosphate dehydrogenase 2; inosine 5'-phosphate dehydrogenase 2; inosine monophosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; inosine-5'-monophosphate dehydrogenase 2; inosine-5'-monophosphate dehydrogenase 2 {ECO:0000255 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23918, 301005, 3615 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6670 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9PU28, P12268, P24547 | |
| IMPDH2 | |
| A synthesized peptide derived from human IMPDH2 (120-175AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction