missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMMP2L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | IMMP2L |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
IMMP2L Polyclonal specifically detects IMMP2L in Human samples. It is validated for Western Blot.Specifications
| IMMP2L | |
| Polyclonal | |
| Rabbit | |
| NP_115938 | |
| 83943 | |
| Synthetic peptide directed towards the C terminal of human IMMP2LThe immunogen for this antibody is IMMP2L. Peptide sequence GHSFDSNSFGPVSLGLLHAHATHILWPPERWQKLESVLPPERLPVQREEE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| EC 3.4.21, EC 3.4.21.-, IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae), IMP2 inner mitochondrial membrane protease-like (S. cerevisiae), IMP2inner mitochondrial membrane peptidase 2 like, IMP2-LIKE, IMP2-like protein, mitochondrial inner membrane protease subunit 2 | |
| IMMP2L | |
| IgG | |
| 20 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title