missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMMP2L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79839
This item is not returnable.
View return policy
Description
IMMP2L Polyclonal specifically detects IMMP2L in Human samples. It is validated for Western Blot.
Specifications
| IMMP2L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.4.21, EC 3.4.21.-, IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae), IMP2 inner mitochondrial membrane protease-like (S. cerevisiae), IMP2inner mitochondrial membrane peptidase 2 like, IMP2-LIKE, IMP2-like protein, mitochondrial inner membrane protease subunit 2 | |
| Rabbit | |
| 20 kDa | |
| 100 μL | |
| Signal Transduction | |
| 83943 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_115938 | |
| IMMP2L | |
| Synthetic peptide directed towards the C terminal of human IMMP2LThe immunogen for this antibody is IMMP2L. Peptide sequence GHSFDSNSFGPVSLGLLHAHATHILWPPERWQKLESVLPPERLPVQREEE. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Horse: 86%; Pig: 85%; Guinea pig: 85%; Bovine: 79% Chicken: 76%. | |
| Human, Bovine, Equine, Guinea Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction