Learn More
Merck Immobilon™-E Transfer Membrane
Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step.
Brand: Merck IEVH09120
177.68 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Features:
- 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
- Saves time and reduces waste by eliminating the alcohol pre-wet step
- Higher protein retention delivers stronger signal than most nitrocellulose membranes
Specifications
Easily wetted with water | |
White | |
Plain | |
Sheet | |
0.45 μm |
Western Blotting | |
IEVH | |
10 Sheets | |
PVDF | |
9 x 12 cm |