missing translation for 'onlineSavingsMsg'
Learn More

Iminodibenzyl, 97%

Product Code. p-148156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10645932 25 g Glass bottle 25g
10316760 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10645932 Supplier Thermo Scientific Acros Supplier No. 122070250
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 494-19-9 | C14H13N | 195.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-19-9
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00005070
InChI Key ZSMRRZONCYIFNB-UHFFFAOYSA-N
Synonym iminodibenzyl, 10,11-dihydro-5h-dibenzo b,f azepine, 10,11-dihydro-5h-dibenz b,f azepine, iminobibenzyl, 2,2'-iminodibenzyl, 2,2'-iminobibenzyl, iminodibenyl, 5h-dibenz b,f azepine, 10,11-dihydro, unii-262bx7oe3u, 10,11-dihydro-5-dibenz b,f azepine
PubChem CID 10308
IUPAC Name 6,11-dihydro-5H-benzo[b][1]benzazepine
SMILES C1CC2=CC=CC=C2NC2=CC=CC=C12

Specifications

Melting Point 105°C to 108°C
Color Beige to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 195.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Iminodibenzyl, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Iminodibenzyl
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.