missing translation for 'onlineSavingsMsg'
Learn More

Iminodibenzyl, 97%

Product Code. 11489476
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489476 100 g 100g
11479476 25 g 25g
11499476 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489476 Supplier Thermo Scientific Alfa Aesar Supplier No. A19204.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 494-19-9 | C14H13N | 195.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-19-9
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00005070
InChI Key ZSMRRZONCYIFNB-UHFFFAOYSA-N
Synonym iminodibenzyl, 10,11-dihydro-5h-dibenzo b,f azepine, 10,11-dihydro-5h-dibenz b,f azepine, iminobibenzyl, 2,2'-iminodibenzyl, 2,2'-iminobibenzyl, iminodibenyl, 5h-dibenz b,f azepine, 10,11-dihydro, unii-262bx7oe3u, 10,11-dihydro-5-dibenz b,f azepine
PubChem CID 10308
SMILES C1CC2=CC=CC=C2NC2=CC=CC=C12

Specifications

Melting Point 105°C to 108°C
Odor Odorless
Quantity 100 g
Beilstein 152732
IUPAC Name 6,11-dihydro-5H-benzo[b][1]benzazepine
Formula Weight 195.27
Percent Purity 97%
Chemical Name or Material Iminodibenzyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.