missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-15N2, TRC

Product Code. 30405670
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405670 10 mg 10mg
30389543 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405670 Supplier Toronto Research Chemicals Supplier No. TRCI35020210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74362-46-2
Molecular Formula C3H415N2
Molecular Weight (g/mol) 70.06
Synonym 1H-Imidazole-1,3-15N2 (9CI, ACI), 15N2-Imidazole, 1H-Imidazole-15N2, Imidazole-15N2
SMILES c1c[15nH]c[15n]1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)/i4+1,5+1
Chemical Name or Material Imidazole-15N2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.