CAS RN 74362-46-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Imidazole-15N2, TRC
CAS: 74362-46-2 Molecular Formula: C3H415N2 Molecular Weight (g/mol): 70.06 Synonym: 1H-Imidazole-1,3-15N2 (9CI, ACI),15N2-Imidazole,1H-Imidazole-15N2,Imidazole-15N2 SMILES: c1c[15nH]c[15n]1