missing translation for 'onlineSavingsMsg'
Learn More

Imazapic, TRC

Product Code. 30407209
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407209 1 g -
30388859 100 mg -
30400270 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407209 Supplier Toronto Research Chemicals Supplier No. TRCI2686351G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104098-48-8
Molecular Formula C14 H17 N3 O3
Molecular Weight (g/mol) 275.3
Synonym 3-Pyridinecarboxylic acid, 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-methyl-, 3-Pyridinecarboxylic acid, 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-methyl-, (±)-, (±)-Imazapic, 2-(4-Isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)-5-methylnicotinic acid, 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-methyl-3-pyridinecarboxylic acid, 5-Methyl-2-(4-isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)nicotinic acid, AC 263222, Cadre, Flame, Flame (herbicide), Imazameth, Imazamethapyr, Imazapic, Imazmethapyr, Oroban
IUPAC Name 5-methyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)pyridine-3-carboxylic acid
SMILES CC(C)C1(C)N=C(NC1=O)c2ncc(C)cc2C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H17N3O3/c1-7(2)14(4)13(20)16-11(17-14)10-9(12(18)19)5-8(3)6-15-10/h5-7H,1-4H3,(H,18,19)(H,16,17,20)
Formula Weight 275.127
Chemical Name or Material Imazapic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.