missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT6/CD85e/LILRA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-98549
This item is not returnable.
View return policy
Description
ILT6/CD85e/LILRA3 Polyclonal specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for Western Blot.
Specifications
| ILT6/CD85e/LILRA3 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31 | |
| Rabbit | |
| 45 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_006856 | |
| LILRA3 | |
| The immunogen for this antibody is LILRA3 - C-terminal region. Peptide sequence AADSPLRLKSKRQSHKYQAEFPMSPVTSAHAGTYRCYGSLSSNPYLLTHP. | |
| Affinity purified | |
| RUO | |
| 11026 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction