missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT2/CD85j/LILRB1 Antibody (VMP55) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £338.00
Specifications
| Antigen | ILT2/CD85j/LILRB1 |
|---|---|
| Clone | VMP55 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692819
|
Novus Biologicals
NBP2-50475-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603828
|
Novus Biologicals
NBP2-50475 |
0.1 mL |
£338.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT2/CD85j/LILRB1 Monoclonal antibody specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOFSpecifications
| ILT2/CD85j/LILRB1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| 10859 | |
| IgG1 κ | |
| Protein A purified |
| VMP55 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7 | |
| Hairy cell leukaemia cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title