missing translation for 'onlineSavingsMsg'
Learn More

ILT2/CD85j/LILRB1 Antibody (VMP55) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50475

 View more versions of this product

Product Code. 18603828

  • £338.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ILT2/CD85j/LILRB1 Monoclonal antibody specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ILT2/CD85j/LILRB1
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7
Hairy cell leukaemia cells
0.1 mL
Immunology
10859
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
VMP55
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.