missing translation for 'onlineSavingsMsg'
Learn More

IL22RA2 Mouse anti-Human, Clone: 5G11A9, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen IL22RA2
Clone 5G11A9
Concentration 1.35 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16807845
View Documents
Proteintech
66190-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16897835
View Documents
Proteintech
66190-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.
Specifications

Specifications

IL22RA2
1.35 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CRF2 10, CRF2 S1, CRF2X, IL 22 receptor subunit alpha 2, IL 22BP, IL 22R alpha 2, IL 22RA2, IL22BP, IL22RA2, Interleukin 22 binding protein, ZcytoR16
IL22RA2
IgG2a
Protein A
Antibody
5G11A9
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
Q969J5
116379
IL22RA2 Fusion Protein Ag18682
Primary
-20°C
IL22RA2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.