missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL22RA2 Mouse anti-Human, Clone: 5G11A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66190-1-IG-150UL
This item is not returnable.
View return policy
Description
IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.Specifications
| IL22RA2 | |
| Monoclonal | |
| 1.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969J5 | |
| IL22RA2 | |
| IL22RA2 Fusion Protein Ag18682 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 5G11A9 | |
| Unconjugated | |
| IL22RA2 | |
| CRF2 10, CRF2 S1, CRF2X, IL 22 receptor subunit alpha 2, IL 22BP, IL 22R alpha 2, IL 22RA2, IL22BP, IL22RA2, Interleukin 22 binding protein, ZcytoR16 | |
| Mouse | |
| Protein A | |
| RUO | |
| 116379 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction